Learn More
Metro™ HD Super Stainless Lab Worktable, Stationary
Keeping things where the belong in a lab is important to all staff. The Metro HD Super Stainless Lab Worktable is stationary and features an open base that makes it easy to clean. Its stable surface and base construction makes it a choice fit for nearly any lab.
Marca:  Metro™ WT367US
Dettagli aggiuntivi : Peso : 38.28000kg
Codice prodotto. 15977331
  • 2205.00€ / Pezzo
Spedizione stimata: 07-02-2025
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • 34 in. high (86.4cm) seamless, 14-gauge stainless-steel work surface with rounded edges
  • Support structure virtually eliminates “drumming”
  • Posts with adjustable feet compensate for slight floor irregularities
  • Leveling foot on each post
Order Info
Shipped knocked down. Assemble easily without tools. Other sizes available; call your Fisher Safety Customer Service Representative at 1-800-772-6733.
Specifica
Specifica
72 in.
Open with side and rear crossrails
Ideal for lab
34 in.
36 in.
106 lb.
182.5 cm
Stainless Steel
Stationary Table
72 x 36 x 34 in. (91.4 x 91.4 x 86.36 cm)
86.36 cm
Bottom 3-sided frame (SS)
48 kg
91.4 cm
NSF