Learn More
Metro™ Super Erecta™ Top Track Mobile Unit Kits
Codice prodotto. 15967301
Click to view available options
Materiale:
Stainless Steel
Steel
Larghezza (sistema imperiale) ripiano:
18 in.
21 in.
24 in.
Finitura:
Chrome
Metroseal 3 with Microban™
Metroseal Green Epoxy
None
Dimensione della confezione:
Pezzo
Codice prodotto. 15967301
Marca: Metro™ TTM24K3
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Includes
Posts, casters/caster channels, donut bumpers and roller bearing assemblies
High-density, easy access storage.
Top-Track storage systems increase storage capacity and ease of access with overhead guide tracks. This design allows for open aisles eliminating hard to reach spaces, an interchangeable/modular storage system and easier cleaning.
  • Mobile units move along anchored track allowing easy access to all shelves
MetroSeal 3 Finish
Is a multi-layer corrosion resistant finish consisting of a proprietary cross linked thermoset epoxy enhanced with Microban antimicrobial protection over a zinc chromate coating. Metroseal 3 with Microban provides protection from bacteria, mold, mildew, and fungi build-up that may cause odor, stains, and product degradation, while displaying excellent abrasion, gouge, and chemical resistance properties.
Order Info
Shelves are ordered separately. One kit required per mobile unit.
Specifica
Product Type Mobile Unit Kit
Finitura Metroseal Green Epoxy
Da utilizzare con (apparecchiatura) Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ Top-Track™ storage system
Include Posts, casters/channel caster, donut bumpers and roller bearing assemblies, hardware, split sleeves
Peso (sistema imperiale) 34 lb.
Peso (sistema metrico) 15.3 kg
Larghezza (sistema imperiale) ripiano 24 in.
Larghezza (sistema imperiale) 26.25 in.
Larghezza (sistema metrico) ripiano 61 cm
Larghezza (sistema metrico) 66.7 cm
Materiale Steel