Learn More
Metro™ Accessorio per scaffali metallici Super Erecta™, rotelle
Codice prodotto. p-63249
Click to view available options
Capacità portata carico (sistema metrico):
68 kg
90 kg
111 kg
135 kg
Product Type:
Brake Stem Caster
High-temperature Autoclave Brake Stem Caster
High-temperature Autoclave Swivel Stem Caster
Rigid Stem Caster
Swivel Caster
Swivel Stem Caster
Swiveled Stem Caster
Materiale:
Antimicrobial Polymer
Polymer
Stainless Steel
Steel
Passo della ruota:
High Modulus Donut
High-temp Nylon
High-temp Phenolic
Polyurethane, Flat
Resilient, Flat
Dimensione della confezione:
Pezzo
The total weight of equipment and load should not exceed three times the load rating per caster.
Codice prodotto. 15906721
Marca: Metro™ 5MDGSA
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
The total weight of equipment and load should not exceed three times the load rating per caster.
Includes
Donut bumpers.
Choose from various types of casters to match your corrosion resistance needs
Caster choices include standard stem, polymer, stainless steel cart-washable and high-temperature autoclavable. Both polymer and stainless casters provide superior corrosion resistance, performance and enhanced durability. Casters available in swivel, swivel brake or rigid styles.
  • Rigid stem casters are ordered two per unit for improved tracking and handling
  • Rigid caster channels are provided with each two rigid stem casters and is based on shelf width
Polymer Casters
  • Polymer casters offer corrosion resistance and enhanced durability
  • All-polymer horn, stainless steel axle and hardware
  • Polyurethane wheel
  • Sleeve-style axle bearing
  • Optional toe-operated brake mechanism
Stainless Steel Cart-Washable Casters
  • Grease sealed with zerk fittings in swivel and axle
High-temperature Autoclave Casters
  • Temperature rated for up to 300°F (149°C)
Order Info
Shelf width must be known when ordering rigid casters. Brakes are foot-operated.
Specifica
Product Type Swiveled Stem Caster
Autoclavabile Not Autoclavable
Diametro (sistema imperiale) 5 in.
Diametro (sistema metrico) 12.7 cm
Da utilizzare con (apparecchiatura) Metro™ Super Erecta™ wire shelving
Capacità supporto carico (sistema imperiale) 150 lb.
Capacità portata carico (sistema metrico) 68 kg
Intervallo di temperatura (sistema imperiale) -40 to 180°F
Temperature Range (Metric) -40 to 82°C
Passo della ruota High Modulus Donut
Con ruote Yes
Materiale Stainless Steel