Learn More
METTLER TOLEDO™ Portable pH/Conductivity/Dissolved Oxygen/Ion Meters
Codice prodotto. 11918011 Sfoglia Tutto Mettler Toledo Prodotti
Click to view available options
Modello:
S9 DO Meter
Seven2Go Pro S8
Seven2Go S2
Seven2Go S3
Seven2Go S4
SevenGo Duo SG23
SevenGo Duo SG68
SevenGo Duo SG78
SevenGo Duo SG98
pH, ORP
Canali:
1
2
Product Type:
Conductivity Meter
DO Meter
DO Portable Meter
Kit da campo
Kit elettrodi
Meter with BOD Kit
Meter with Bioethanol Kit
Meter with Electrode Kit
Meter with Field Kit
Meter with Field Kit, 2m Cable
Meter with Field Kit, 5m Cable
Meter with Food Kit
Meter with Light Kit
Meter with Standard Kit
Misuratore per pH/ mV e ioni Seven2Go Pro S8
Misuratore per pH/ mV e ioni Seven2Go Pro S8: Kit Biotech
Misuratore per pH/ mV e ioni Seven2Go Pro S8: Kit da campo
Misuratore per pH/ mV e ioni Seven2Go Pro S8: Kit fluoruro
Solo misuratore (SG23)
pH/Ion/Cond Meter
pH/Ion/DO Meter
pH/Ion/DO/ORP Meter
pH/mV Meter
Dimensione della confezione:
1 set
Pezzo
Codice prodotto. 11918011
Marca: METTLER TOLEDO™ 51302664
Water and dust proof, IP67 rated meter for portable or field measurements. Portable meters compatible with pH, conductivity, dissolved oxygen, and ion selective electrodes.
  • Dust proof
  • Water proof
  • IP67 Rated
  • pH, Redox/ORP, conductivity, dissolved oxygen and ISE measuring capability (model dependent)
  • Compatible with a wide range of IP67-rated electrodes for any application
  • Compatible with our ISM electrodes that capture and save calibration data which is then automatically recognized by the meter once connected
  • Data storage available; capacity dependent on model
  • Export data to a printer or Excel file for Seven2Go Pro only
  • EasyDirect software available for data transfer (model dependent) for Seven2Go Pro only
  • Rubber holster available for single-handed measurements
  • uGo carrying and protecting case included in selective kits (see below for complete list); available for all portable meters
TRUSTED_SUSTAINABILITY
Specifica
Uscite IR to printer or PC via RS232 or USB
Precisione (mV) ±0.1mV
Precisione (pH) ±
Tipo di batteria 4 x AA
Punti di calibrazione 5
Certificazione/Conformità ISO 9001:2000
Canali 2
Tipo di display Backlit Graphic LCD
Protezione IP IP67
Measurement Modes pH, Dissolved Oxygen
Intervallo (mV) -1999 to 1999 mV
Intervallo (pH) -2 to 19.999
Risoluzione (pH) 0.001
Temperature Range (Metric) -5°C to +130°C
Include InLab OptiOx-5m Dissolved oxygen/temperature electrode; InLab Expert Go-5m-ISM pH/temperature electrode
Ingressi InLab Expert Go-5m-ISM pH/temperature electrode
Modello SevenGo Duo SG98
Product Type Meter with Field Kit, 5m Cable
Risoluzione (mV) 0.1mV
Precisione termica ±0.2°C
Temperatura (sistema imperiale) operativa 32°F to 104°F
Temperatura (sistema metrico) operativa 0°C to 40°C
Risoluzione temperatura 0.1°C
Dimensioni (L x P x A) 8.66 x 3.54 x 1.77 in. (22 x 9 x 4.5cm)
Lunghezza (sistema imperiale) 8.66 in.
Lunghezza (sistema metrico) 22 cm
Larghezza (sistema imperiale) 3.54 in.
Larghezza (sistema metrico) 9 cm
Altezza (sistema imperiale) 1.77 in.
Altezza (sistema metrico) 4.5 cm