Learn More
Greiner Bio-One Micropiastre ELISA da 96 pozzetti in polistirene con fondo a F Microlon™
In polistirene trasparente per garantire un’ottima visibilità. Le micropiastre ELISA Greiner BioOne™ Microlon™ in polistirolo a 96 pozzetti sono testate in un immunodosaggio colorimetrico per applicazioni di ricerca immunologica.
Marca: Greiner Bio-One 655061
Dettagli aggiuntivi : Peso : 1.89000kg
Descrizione
- Free of detectable DNase, RNase, human DNA
- Non-Pyrogenic
- Manufactured under DIN ISO 9001 guidelines
- Tracebility from production through a defined LOT number system
- Manufactured from crystal clear polystyrene
- The footprint is compatible with automated systems
- Well to well spacing : 9 mm
- Alphanumeric well coding
- Suitable for transmission measurements
- Specifically treated to provide an incresed protein binding
- Well profile: F-bottom / standard (flat bottom)
Specifica
High Binding | |
Without Lid | |
Trasparente | |
96 | |
Flat Bottom (F-well) | |
Standard Well |
Standard | |
8 x 12 | |
Polistirene | |
Microlon 600 | |
Confezione da 40 |