Learn More
Thermo Scientific™ MicroTest™ Universal Plus Kits
Maintain organism viability with kits including transport media and two traditional-tipped swabs plus an envelope with both traditional and micro-tipped polyester swabs.
438.00€
Specifica
Product Type | Universal Kit |
---|---|
Formato | Tube |
SwabConfiguration | Dual Applicator (1 Traditional, 1 Microtip Swab) |
Compatibilità | Culture, Shell vial, Freeze-thaw cycles, Culture maintenance, ELISA, PCR, DNA probe technology |
Quantità | 100 Kits/Pk. |
Codice del prodotto | Marca | Descrizione | Quantità | Volume (sistema metrico) | Da utilizzare con (applicazione) | Prezzo | Quantità e disponibilità | ||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Descrizione | Quantità | Volume (sistema metrico) | Da utilizzare con (applicazione) | Prezzo | Quantità e disponibilità | ||
15219467
|
Thermo Scientific™
R12576 |
M4RT Universal Plus Kit | 100 Kits/Pk. | 3 mL | Chlamydiae Viruses |
438.00€
100 confezioni |
Spedizione stimata: 29-08-2024
Eseguire il login per visualizzare lo stock disponibile |
||
Descrizione
Maintain organism viability and counts for verification studies, quality control procedures, and repeat testing of viruses chlamydiae, mycoplasma and ureaplasma with quality media, proven collection and transport tools, and safe, reliable packaging. Thermo Scientific™ MicroTest™ Universal Plus Kits come in two media formulations and include two plastic shaft traditional-tipped polyester swabs plus an additional envelope with one plastic shaft traditional-tipped polyester swab and one stainless steel shaft micro-tipped polyester swab and a 6 x 9 in. zip-seal bag for safe transport.
Designed for the collection, transportation, maintenance and long-term storage of viral specimens, chlamydiae, mycoplasma and ureaplasma, Thermo Scientific MicroTest products are top quality. From formulation to packaging, MicroTest products maintain the viability of organisms through freeze-thaw cycles while inhibiting antimicrobial contaminants and promoting safety.MicroTest™ M4™ contains gelatin, vancomycin, amphotericin B, and colistin for the transport of viruses, Chlamydiae, Ureaplasmas, and Mycoplasmas.
MicroTest™ M4RT™ contains gelatin, gentamicin, and amphotericin B for the transport of viruses and Chlamydiae.
MicroTest Universal Plus Kits come with 1.5mL or 3mL of liquid medium in a 15mL conical tube with 3 glass beads, an envelope with two plastic shaft traditional-tipped polyester swabs plus an envelope with one plastic shaft, traditional-tipped polyester swab and one stainless steel shaft micro-tipped polyester swab, and a 6 x 9 in. zip-seal bag.
Keep your lab moving in the right direction with a dependable set of solutions for transport, holding, maintenance and long-term culture storage with the MicroTest family of collection and transport media.
Trusted reliable results
- Stringent quality control, including cellular toxicity on each lot number
- Patented formulations with stabilizing proteins, buffers, and specific antibiotics
- Provides compatible transport for cell culture, enzyme immunoassays, and molecular diagnostic tests
- Maintains organism viability and counts for better, more reliable results during validation/verification studies, quality control procedures, and repeat testing.
- 1.5mL and 3.0mL fill volumes allow multiple tests from one specimen
- Individual transport media tubes or complete kits offer flexibility and versatility
- Tough, break-resistant tubes with secure, color-coded screw caps prevent leaking and exposure to infectious agents
- Kits include leak-resistant polypropylene bag for safe sample transport
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifica
Universal Kit | |
Dual Applicator (1 Traditional, 1 Microtip Swab) | |
100 Kits/Pk. | |
Traditional and Microtipped |
Tube | |
Culture, Shell vial, Freeze-thaw cycles, Culture maintenance, ELISA, PCR, DNA probe technology | |
Plastic, Stainless Steel | |
3 mL |