Learn More
Kinematica™ Mixer da laboratorio MB 800 Microtron™
A laboratory mixer well-suited for intense mixing, dispersing, homogenizing, emulsifying and mincing operations for cuttable materials, making samples easy to work with for analysis and quality control.
Marca: Kinematica™ 36010003
Dettagli aggiuntivi : Peso : 0.80000kg
Descrizione
- Smooth rotational speed adjustment with integrated on/off switch
- Load compensated rotational speed adjustment
- Switching of mixing attachment and protection cover without tools
- Operation only possible with security cover and lid in place
- Variety of sample container designs
- Vibration dampening quick coupling
- Very quiet motor with integrated ventilation cooling
- Integrated load, heat and blockage protection
- Production of very fine particle sizes in both liquid and dry mediums - precise analyses require homogenous samples
- Efficient sample preparation through high rotational speeds
- Flexibility with various sample volumes with different mixing attachments
- Durable, robust, long-lasting design
Specifica
Mixer da laboratorio | |
230 V | |
800 W | |
245 mm | |
MB 800 with EU Power Cable | |
Confezione da 1 |
1000 to 17,000 rpm | |
60 Hz | |
2 to 4 L | |
Per miscelazione, dispersione, omogeneizzazione, emulsificazione e frantumazione di alimenti | |
79 dBA at 17,000 rpm | |
5.3 kg |