Learn More
Cytiva Mini Dialysis Kits
Sfoglia Tutto Cytiva Prodotti
Click to view available options
MWCO:
1 kDa
8 kDa
Capacità (sistema metrico):
250 μL
2 mL
Dimensione della confezione:
1 set
250µL
Pezzo
Description
Includes
50 tubes with 50 dialysis and 50 standard caps, plus six floats
- Especially well-suited for desalting protein samples prior to analysis by isoelectric focusing (IEF) and 2-D electrophoresis
- Uses disposable dialysis tubes with dialysis membrane incorporated into caps
- Conical tube bottom maximizes sample recovery
- Process complete in as little as several hours
- Choice of molecular weight cut-off and tube size
Specifications
Specifications
| Product Type | Mini Dialysis Kit |
| MWCO | 8 kDa |
| Capacità (sistema metrico) | 250 μL |
| Contenuto kit | 50 x tube with dialysis cap, 50 x standard cap, 6 x float |
| Quantità | Each |
| Da utilizzare con (applicazione) | Offers a simple solution to the handling problems of small volume dialysis, the dialysis tubes in the kit each have a cap that incorporates a small disk of dialysis membrane |