Learn More
3M™ Mini Flip-Top Vial
Leak-resistant plastic cylinder with a flip-open lid that seals securely for safe shipping and handling.
Marca: 3M™ 7000030317
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Hinged lid can be opened with one hand and stays open for easy sample additions.
- Decontaminated through gamma irradiation and ready for use with no additional blending required.
- Flat bottom tube stands on its own, no holder required.
- Leak resistant vial helps prevent leakage during shipping.
- 2 year shelf life from date of manufacture provides long-term readiness.
Specifica
Food and Beverage, Environmental Test | |
9 mL Maximum Recovery Diluent | |
100/Cs. |