Learn More
Erlab™ Molecode F
Erlab™ Molecode F is a saturation detection alarm for broad spectrum filters.
Marca: Erlab™ I3099000002
761.92 EUR valido fino al 2024-09-17
Usa il codice promo "21690" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 1.00000kg
Descrizione
- Equipment required by class I of the AFNOR NF X 15-211: 2009 standard
- A cell located in the sampling chamber allows detection Automatic saturation of the filter by formaldehyde
- A second cell is placed in contact with the environment of the laboratory indicates a possible pollution of the ambient air
Specifica
Captair Smart Fume Hoods | |
Saturation Detection Alarm | |
Detecting Formaldehyde |