Learn More
Invitrogen™ Mouse IFNa ProQuantum Immunoassay Kit
The ProQuantum Mouse IFNa Immunoassay Kit is designed to provide quantitative measurements of mouse IFNa in small sample volumes.
Marca: Invitrogen™ A46736
Descrizione
Utilizing proximity-based amplification technology, the assay combines the analyte specificity of high-affinity antibody-antigen binding with the signal detection and amplification capabilities of real-time PCR to achieve a simple yet powerful next-generation protein quantitation platform. A user-friendly workflow combined with intuitive software for analytics enables sample-to-answer in just 2 hours.
- High sensitivity-detect low levels of protein with greater sensitivity than traditional methods
- Broad dynamic range-≥5 logarithmic units, minimizing sample dilutions to ensure they fall within the range
- Small sample consumption-use 2–5 μL of sample (compared to 75 μL for triplicate wells with other methods)
- Fast, easy workflow-2 hours from sample to answer, with no wash steps
- No proprietary instrument to purchase-runs on any real-time PCR instrument
Specifica
alpha-2 A interferon; Gm12597; ifa; Ifa1; Ifa2; IFL; IFN; IFN alpha; IFN a; IFN1@; IFNA; IFNA@; IFNA1; IFNA13; Ifna15; IFNA2; IFNA2A; IFNA2B; IFNA2C; IFNA3; IFNA6; Ifnaa; IFNAalphaIFN-alpha-1/13; IFNalpha; IFN-alpha; IFN-alpha 1 b; IFN-alpha1; IFN-alpha-1; IFN-alpha-1/13; IFN-alpha-2; IFN-alpha-6; IFN-alphaA; IFN-alphaD; IFN-gamma; IFNa; INFA2; interferon; interferon alpha; interferon alpha 1; interferon alpha 15; interferon alpha 1 b; interferon alpha 2; interferon alpha 2 A; interferon alpha 2 b; interferon alpha A; interferon alpha family gene 1; interferon alpha family, gene 2; interferon alpha protein; interferon alpha-1; interferon alpha-1 precursor; Interferon alpha-1/13; interferon alpha-2; Interferon alpha-A; Interferon alpha-D; interferon type A1/A2; interferon type A3; interferon, alpha 1; interferon, alpha 2; interferon-alpha; interferon-alpha 1; interferon-alpha1; interferon-alpha-6; leIF A; LeIF D; MGC138207; MGC138505; MGC138507; MuIFN-alpha-A; novel interferon alpha (Ifna) family member; OTTMUSG00000007655 | |
<1 pg/mL | |
qPCR Instruments | |
IFNA, IFNA1, Ifna15, IFNA2 | |
96 Tests | |
Serum, 2-5 μL; Plasma, 2-5 μL; Supernatant, 2-5 μL | |
IFN alpha | |
2 hr |
1.6 to 25,000 pg/mL | |
ELISA | |
P01573, P01572 | |
Assay Dilution Buffer Antibody-Conjugate A Antibody-Conjugate B Antibody-Conjugate Dilution Buffer Standard Master Mix Ligase | |
Research Use Only | |
-20°C | |
30 min |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.