missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse IL-22 ProQuantum Immunoassay Kit
ELISA
Marca: Invitrogen™ A47434
634.12 EUR valido fino al 2025-03-28
Usa il codice promo "24107" per avere il tuo prezzo promozionale.
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Utilizing proximity-based amplification technology, the assay combines the analyte specificity of high-affinity antibody-antigen binding with the signal detection and amplification capabilities of real-time PCR to achieve a simple yet powerful next-generation protein quantitation platform. A user-friendly workflow combined with intuitive software for analytics enables sample-to-answer in just 2 hours.
- High sensitivity-detect low levels of protein with greater sensitivity than traditional methods
- Broad dynamic range-≥5 logarithmic units, minimizing sample dilutions to ensure they fall within the range
- Small sample consumption-use 2–5 μL of sample (compared to 75 μL for triplicate wells with other methods)
- Fast, easy workflow-2 hours from sample to answer, with no wash steps
- No proprietary instrument to purchase-runs on any real-time PCR instrument

Specifica
Cytokine Zcyto18; EGK | |
<0.3 pg/mL | |
ProQuantum | |
qPCR Instruments | |
IL22 | |
96 Tests | |
Plasma 2-5 μL, Serum 2-5 μL, Supernatant 2-5 μL | |
IL-22 | |
2 hrs |
0.32 to 25,000 pg/mL | |
FAM™ | |
Plasma, Serum, Supernatant | |
50929 | |
Assay Dilution Buffer, Antibody-Conjugate A, Antibody-Conjugate B, Antibody-Conjugate Dilution Buffer, Standard, Master Mix, Ligase | |
RUO | |
-20°C | |
30 mins |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto