Learn More
R&D Systems™ Mouse Oncostatin M (OSM) Quantikine ELISA Kit
a 4.5 hour solid phase ELISA designed to measure mouse OSM levels in cell culture supernates, serum, and plasma.
Marca: R&D Systems™ MSM00
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The Mouse Oncostatin M (OSM) Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse Oncostatin M/OSM in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul).Specifica
7.81 - 500pg/mL | |
HRP | |
Natural and Recombinant Mouse OSM | |
18413 | |
Store the unopened product at 2° to 8°C. Do not use past expiration date. |
4.7pg/mL | |
Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum | |
ELISA | |
Cell Culture Supernates (50uL),Serum (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL) |