missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Sonic Hedgehog/Shh N-Terminus Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
Marca: R&D Systems MAB4641-SP
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Sonic Hedgehog/Shh Monoclonal specifically detects Sonic Hedgehog/Shh in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Neutralization, ELISA Capture (Matched Antibody Pair).
Specifica
| Sonic Hedgehog/Shh | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| HHG1, HHG-1, HLP3, HPE3, MCOPCB5, MCOPCB5sonic hedgehog (Drosophila) homolog, Shh, ShhNC, SMMCI, SMMCIsonic hedgehog homolog (Drosophila), sonic hedgehog, sonic hedgehog homolog, sonic hedgehog protein, TPT, TPTPS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6469 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, Immunohistochemistry, Neutralization, ELISA | |
| 171018 | |
| Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL, Neutralization 5-15 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Q62226 | |
| SHH | |
| E. coli-derived recombinant mouse Sonic Hedgehog Cys25-Gly198 (Lys122Arg) Accession # Q62226 | |
| 25 μg | |
| Primary | |
| Detects mouse Sonic Hedgehog in ELISAs. In Western blots, approximately 50% cross-reactivity with recombinant mouse (rm) Dhh (N-terminus; aa 23-198) is observed and no cross-reactivity with rmIhh (N-terminus; aa 66-240), rmDhh (C-terminus; aa 199-396), or rmShh (C-terminus; aa 199-437) is observed. | |
| Mouse | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto