Learn More
R&D Systems™ Mouse XL ICAM-1/CD54, Luminex™ Performance Assay
Codice prodotto. 18422470
Click to view available options
Quantità:
1 Kit
Dimensione della confezione:
1 set
Codice prodotto. 18422470
Marca: R&D Systems™ LMXL796
Luminex bead-based multiplex assays are designed to provide accurate, reproducible results for every target analyte.
TRUSTED_SUSTAINABILITY
Specifica
Quantità 1 Kit
Specie Mouse
ID gene (immissione) 3383
Sinonimo BB2, CD54, CD54 antigen, cell surface glycoprotein P3.58, human rhinovirus receptor, ICAM1, ICAM-1, intercellular adhesion molecule 1, intercellular adhesion molecule 1 (CD54), human rhinovirus receptor, Major group rhinovirus receptor, P3.58
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.