Learn More
MP Biomedicals™ 1/4 in. Ceramic Spheres
Codice prodotto. 11412400 Sfoglia Tutto MP Biomedicals Prodotti
Click to view available options
Quantità:
100 Pk.
1000 Pk.
50 Pk.
500 Pk.
Dimensione della confezione:
pezzi 100
pezzi 1000
pezzi 50
pezzi 500
Codice prodotto. 11412400
Marca: MP Biomedicals™ 116540412
Use to break hard/brittle samples. Specialty applications for the 1/4 in. Ceramic Spheres include RNA extraction from difficult samples where lysis by impaction only is preferable with minimal amounts of mechanical shearing.
  • Zirconium Oxide-coated ceramic grinding spheres
  • Chemically inert
  • Component of Lysing Matrix A and M tubes
TRUSTED_SUSTAINABILITY
Specifica
Tipo di profilo Zirconium oxide coated ceramic
Content And Storage Room Temperature
Tipo di campione Bone, Heart, Lung, Seeds, Skeletal Muscle, Spores
Da utilizzare con (applicazione) Break hard/brittle samples
Product Type Bulk Matrix
Quantità 500 Pk.
Volume del campione Unlimited
Sinonimo 1/4 in. Zirconium Oxide Banded Satellite for Grinding
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.