Learn More
MP Biomedicals™ FastDNA™-96 Fecal DNA Kit
The FastDNA-96™ Fecal DNA Kit quickly and efficiently isolates inhibitor-free, PCR-ready genomic DNA from stool samples (i.e. humans, primates, birds, rodents, cattle, canine, feline, porcine, etc.).
Marca: MP Biomedicals™ 119696400
Dettagli aggiuntivi : Peso : 0.68000kg