Learn More
MP Biomedicals™ FastDNA™-96 Fecal DNA Extraction Kit
FastDNA™-96 Fecal DNA Extraction Kit is designed to isolate bacterial, protist, or host DNA from up to 80 mg of wet or dry stool, and even from caecum samples, through a purification process that eliminates PCR inhibitors.
Marca: MP Biomedicals™ 119696400
Dettagli aggiuntivi : Peso : 1.22900kg Transport : UN number : 3316 Chem class : 9 Pack group : II
Descrizione
- Product Overview: The FastDNA-96™ Fecal DNA Kit isolates bacterial, protist, or host DNA from up to 80 mg of wet or dry stool, and even from caecum samples, through a purification process that eliminates PCR inhibitors. Fecal microbes, fungi, parasites, and other stool organisms are first lysed in approximately 60 seconds with the FastPrep-96™ Instrument.
- Applications: The FastDNA-96™ Fecal DNA Kit quickly and efficiently isolates inhibitor-free, PCR-ready genomic DNA from stool samples (i.e. humans, primates, birds, rodents, cattle, canine, feline, porcine, etc.).
- Sample Type: Mammalian Stool (i.e. humans, primates, birds, rodents, cattle, canine, feline, porcine, etc.)
Specifica
Kit | |
2 x 96 Preps | |
1 Ea. |
Mammalian Stool | |
FastDNA-96 Kits |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.