Learn More
MP Biomedicals™ FastDNA™ SPIN Kit for Plant and Animals Tissues
FastDNA™ SPIN Kit for Plant and Animals Tissues is used to isolate DNA from plant and animal tissues.
Marca: MP Biomedicals™ 116540800
Additional Details : Peso : 0.68000kg Transport : UN number : 1760 Chem class : 8 Pack group : III
Requires : FastPrep™ Instrument or alternative high speed disruptor; Microcentrifuge that can freely spin 2.0 mL tubes; Microcentrifuge tubes (2.0 mL and 1.5 mL); Rotator or low-speed vortex
Description
- Product Overview: The FastDNA™ Spin Kit for Plant and Animals Tissues quickly and efficiently isolates high quality genomic DNA from plant and animal tissues using Lysing Matrix D (1.4 mm ceramic beads) for cell lysis and a silica-based spin filter method for the purification process.
- Sample Type: Animal Tissues, Plants, Cultured Cells
Specifications
Animal Tissues, Plants, Cultured Cells | |
100 x 2 mL Tubes Lysing matrix D, 0.25 in. Ceramic Spheres, 100 mL Binding Matrix, 12 mL Concentrated SEWS-M, 25 mL DES, 90 mL CLS-VF, 25 mL PPS, 110 mL CLS-TC, 100 x SPIN Modules, 100 x Catch Tubes | |
FastDNA-24 Kits |
Use to isolate DNA from plant and Animal tissues | |
100 Preps |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.