missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Lysing Matrix C, Bulk
Lysing Matrix C contains 1 mm silica spheres. Lysing Matrix C is used primarily for lysis of yeast, fungi, molds, spores and filamentous algae.
Marca: MP Biomedicals™ 116540432
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Product Overview: 1 mm silica spheres. Found in the FastRNA Pro Red Kit and FastProtein Red Matrix. Lysing Matrix C is used primarily for lysis of yeast and fungi and can also be used for molds, spores and filamentous algae.
- Applications: Lysing Matrix C is used primarily for lysis of yeast and fungi and can also be used for molds, spores and filamentous algae.

Specifica
Store at 2°C to 8°C | |
Yeast, Fungi, Algae | |
Lysing Matrix C | |
500 g |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto