missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MPHOSPH1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA556696
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Immunogen sequence: KVETEEATAC LELKFNQIKA ELAKTKGELI KTKEELKKRE NESDSLIQEL ETSNKKIITQ NQRIKELINI IDQKEDTINE FQNLKSHMEN TFKCNDKADT SSLIINNKLI CNETVEVPKD SKSKICSERK RVNENELQQD EPPAKKGSI Highest antigen sequence identity to the following orthologs: Mouse - 44%, Rat - 53%.
KIF20B is a plus-end-directed motor enzyme that is required for completion of cytokinesis.Specifica
MPHOSPH1 | |
Polyclonal | |
Unconjugated | |
KIF20B | |
33cex; B130024C23; C330014J10Rik; Cancer/testis antigen 90; CT90; Kif20b; Kinesin family member 20B; kinesin-like protein KIF20B; Kinesin-related motor interacting with PIN1; KRMP1; magoo; mitotic kinesin-like protein; mitotic kinesin-related protein; M-phase phosphoprotein 1; MPHOSPH1; MPP1; MPP-1; N-6 kinesin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9585 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96Q89 | |
KIF20B | |
Recombinant protein corresponding to Human MPHOSPH1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only