Learn More
Gilson™ MyPIPETMAN™ Volume Display Windows
For easier identification and convenience, this colored window provides extra pipette customization.
Specifica
Autoclavabile | Autoclavable |
---|---|
Calibrato | No |
Materiale | Polystyrene, Polypropylene |
Sterilità | Non-sterile |
Graduato | No |
Descrizione
- Six available colors
- Easily replace the colored window at your convenience
Specifica
Autoclavable | |
Polystyrene, Polypropylene | |
No | |
Reusable |
No | |
Non-sterile | |
Display Window | |
MyPIPETMAN |