missing translation for 'onlineSavingsMsg'
Learn More

N-(1-Naphthyl)ethylenediamine, dihydrochloride, 98+%, ACS reagent, Thermo Scientific Chemicals

Codice prodotto. 17457532
Click to view available options
Quantità:
5 g
25 g
100 g
500 g
Imballaggio:
Glass bottle
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

N-(1-Naphthyl)ethylenediamine, dihydrochloride, >-98.0% (on dry substance), C12H16Cl2N2, CAS Number-1465-25-4

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 1465-25-4
Numero MDL MFCD00012556
InChI Key MZNYWPRCVDMOJG-UHFFFAOYSA-N
Sinonimo n-1-naphthalenyl-1,2-ethanediamine dihydrochloride, ccris 425, n-1-naphthyl ethylenediamine dihydrochloride, marshall's reagent, bratton-marshall reagent, n-1-naphthylethylenediamine dihydrochloride, 1,2-ethanediamine, n-1-naphthalenyl-, dihydrochloride, n1-naphthalen-1-yl ethane-1,2-diamine dihydrochloride, unii-h734599kjl, 2-1-naphthylamino ethylamine 2hcl
PubChem CID 15106
ChEBI CHEBI:53452

Specifica

Materiale o nome chimico N-(1-Naphthyl)ethylenediamine, dihydrochloride
CAS min. % 98.0
CAS max % 100.0
Informazioni di decomposizione 200°C
Punto di fusione 200.0°C
Colore Beige to White or Pink
Spettro a infrarossi Authentic
Intervallo percentuale saggio 98% min. (on dry substance) (Argentometry)
Imballaggio Glass bottle
Formula molecolare C12H16Cl2N2
Formula lineare C10H7NHCH2CH2NH2·2HCl
Quantità 5 g
Beilstein 12, II, 699
Sensibilità (to sulfanilamide) Passes
Indice di Merck 15, 6491
Informazioni di solubilità Passes test
SMILES [H+].[H+].[Cl-].[Cl-].NCCNC1=C2C=CC=CC2=CC=C1
IUPAC Name dihydrogen N1-(naphthalen-1-yl)ethane-1,2-diamine dichloride
Molecular Weight (g/mol) 259.17
Peso formulazione 259.17
Percent Purity 98+%
Grado ACS Reagent
Forma fisica Powder
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'