Learn More
N-Boc-L-phenylalaninol, 99%, Thermo Scientific Chemicals
Codice prodotto. 11369879
Click to view available options
Quantità:
1 g
5 g
25 g
N-Boc-L-phenylalaninol is employed in the synthesis of biochemically active compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Boc-L-phenylalaninol is employed in the synthesis of biochemically active compounds.
Solubility
Insoluble in water.
Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 66605-57-0
Formula molecolare C14H21NO3
Molecular Weight (g/mol) 251.33
Numero MDL MFCD00076976
InChI Key LDKDMDVMMCXTMO-LBPRGKRZSA-N
Sinonimo n-boc-l-phenylalaninol, boc-l-phenylalaninol, boc-phenylalaninol, s-2-boc-amino-3-phenyl-1-propanol, n-tert-butoxycarbonyl-l-phenylalaninol, tert-butyl n-2s-1-hydroxy-3-phenylpropan-2-yl carbamate, s-tert-butyl 1-hydroxy-3-phenylpropan-2-yl carbamate, s-n-tert-butoxycarbonyl-phenylalaninol, s---2-tert-butoxycarbonylamino-3-phenyl-1-propanol
PubChem CID 2733675
IUPAC Name tert-butyl N-[(2S)-1-hydroxy-3-phenylpropan-2-yl]carbamate
SMILES CC(C)(C)OC(=O)N[C@H](CO)CC1=CC=CC=C1
Specifica
Materiale o nome chimico N-Boc-L-phenylalaninol
Punto di fusione 95°C to 98°C
Beilstein 3652004
Informazioni di solubilità Insoluble in water.
Quantità 25 g
Peso formulazione 251.33
Optical Rotation −27.5° ± 2.5° (c=1 in Methanol)
Percent Purity 99%