Learn More
N-Chloroacetylisonipecotic acid, 98%, Thermo Scientific™
Codice prodotto. 11374885
Click to view available options
Quantità:
1 g
5 g
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 318280-69-2
Formula molecolare C8H12ClNO3
Molecular Weight (g/mol) 205.64
Numero MDL MFCD02093986
InChI Key PEQVHRULPWCLOJ-UHFFFAOYSA-N
Sinonimo n-chloroacetylisonipecotic acid, 1-2-chloroacetyl piperidine-4-carboxylic acid, 1-2-chloroacetyl-4-piperidinecarboxylic acid, acmc-20ak5i, n-chloroacetyl-dl-isonipecotic acid, 1-chloroacetylpiperidine-4-carboxylic acid, 1-chloroacetyl-4-piperidinecarboxylic acid, 1-chloroacetyl piperidine-4-carboxylic acid, 4-piperidinecarboxylic acid, 1-chloroacetyl, 4-piperidinecarboxylic acid, 1-chloroacetyl-9ci
PubChem CID 1991140
IUPAC Name 1-(2-chloroacetyl)piperidine-4-carboxylic acid
SMILES OC(=O)C1CCN(CC1)C(=O)CCl
Specifica
Materiale o nome chimico N-Chloroacetylisonipecotic acid
Punto di fusione 109°C to 111°C
Quantità 1 g
Peso formulazione 205.64
Percent Purity 98%