missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Fmoc-O-benzylphospho-L-threonine, 97%, Thermo Scientific Chemicals
78.50€ - 306.00€
Identificatori chimici
CAS | 175291-56-2 |
---|---|
Formula molecolare | C26H26NO8P |
Molecular Weight (g/mol) | 511.47 |
Numero MDL | MFCD00797870 |
InChI Key | HOFDVXHILSPFNS-OSPHWJPCSA-N |
Sinonimo | fmoc-o-benzylphospho-l-threonine, fmoc-thr po obzl oh-oh, fmoc-thr hpo3bzl-oh, fmoc-l-thr po obzl oh-oh, fmoc-thr po3bzlh-oh, 2s,3r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-benzyloxy hydroxy phosphoryl oxy butanoic acid, n-fmoc-o-benzylphospho-l-threonine, pubchem20602, fmoc-thr po3bzlh-oh hplc, fmoc-o-hydroxy benzyloxy phosphinyl-l-thr-oh |
PubChem CID | 15224585 |
IUPAC Name | (2S,3R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[hydroxy(phenylmethoxy)phosphoryl]oxybutanoic acid |
SMILES | C[C@@H](OP(O)(=O)OCC1=CC=CC=C1)[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
15419438
|
Thermo Scientific Alfa Aesar
H63006.MD |
250 mg |
78.50€
250mg |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
15409438
|
Thermo Scientific Alfa Aesar
H63006.03 |
1 g |
306.00€
1g |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identificatori chimici
175291-56-2 | |
511.47 | |
HOFDVXHILSPFNS-OSPHWJPCSA-N | |
15224585 | |
C[C@@H](OP(O)(=O)OCC1=CC=CC=C1)[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O |
C26H26NO8P | |
MFCD00797870 | |
fmoc-o-benzylphospho-l-threonine, fmoc-thr po obzl oh-oh, fmoc-thr hpo3bzl-oh, fmoc-l-thr po obzl oh-oh, fmoc-thr po3bzlh-oh, 2s,3r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-benzyloxy hydroxy phosphoryl oxy butanoic acid, n-fmoc-o-benzylphospho-l-threonine, pubchem20602, fmoc-thr po3bzlh-oh hplc, fmoc-o-hydroxy benzyloxy phosphinyl-l-thr-oh | |
(2S,3R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[hydroxy(phenylmethoxy)phosphoryl]oxybutanoic acid |
Specifica
N-Fmoc-O-benzylphospho-L-threonine | |
C26H26NO8P | |
7557104 | |
HOFDVXHILSPFNS-OSPHWJPCSA-N | |
(2S,3R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[hydroxy(phenylmethoxy)phosphoryl]oxybutanoic acid | |
15224585 | |
97% | |
Solid |
175291-56-2 | |
MFCD00797870 | |
fmoc-o-benzylphospho-l-threonine, fmoc-thr po obzl oh-oh, fmoc-thr hpo3bzl-oh, fmoc-l-thr po obzl oh-oh, fmoc-thr po3bzlh-oh, 2s,3r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-benzyloxy hydroxy phosphoryl oxy butanoic acid, n-fmoc-o-benzylphospho-l-threonine, pubchem20602, fmoc-thr po3bzlh-oh hplc, fmoc-o-hydroxy benzyloxy phosphinyl-l-thr-oh | |
C[C@@H](OP(O)(=O)OCC1=CC=CC=C1)[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O | |
511.47 | |
511.46 | |
Moisture sensitive |
Sicurezza e movimentazione
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only