missing translation for 'onlineSavingsMsg'
Learn More

n-Heptane, 99+%, for spectroscopy

Codice prodotto. p-216862 Sfoglia Tutto Thermo Scientific Chemicals Prodotti
Click to view available options
Quantità:
500 mL
2.5 L
Imballaggio:
Bottiglia in vetro
EcoSafPak™ Glass Bottle
Dimensione della confezione:
2.50L
500mL
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Normativa: Questo prodotto è soggetto ad accise sui prodotti energetici secondo la direttiva UE 2003/96/CEE.

Codice prodotto. 10559472

Marca: Thermo Scientific Acros 411255000

Richiesta di formato massivo o personalizzato

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 142-82-5 | C7H16 | 100.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-82-5
Formula molecolare C7H16
Molecular Weight (g/mol) 100.21
Numero MDL MFCD00009544
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
Sinonimo n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
PubChem CID 8900
ChEBI CHEBI:43098
IUPAC Name heptane
SMILES CCCCCCC

Specifications

Materiale o nome chimico n-Heptane
Punto di fusione -91.0°C
Absorbance 0.01 max. at 240nm, 0.01 max. at 250nm, 0.01 max. at 260nm, 0.01 max. at 400nm, 0.05 max. at 230nm, 0.15 max. at 220nm, 0.4 max. at 210nm, 1 max. at 197nm, 1 max. at 200nm
CAS min. % 99.0
CAS max % 100.0
Densità 0.6830g/mL
Punti di ebollizione 98.0°C
Quantità 500 mL
Punto d'infiammabilità −4°C
Imballaggio EcoSafPak™ Glass Bottle
Forma fisica Liquido
Indice di rifrazione 1.3850 to 1.3890
Intervallo percentuale saggio 99+%
Residuo dopo l'evaporazione 0.0005 % max.
Gravità specifica 0.683
Formula lineare CH3(CH2)5CH3
Pressione del vapore Vapor Pressure: 48mbar at 20°C
Viscosità 0.4 mPa.s (20°C)
Beilstein 01, 154
Acqua 0.03% Max.
Indice di Merck 15, 4694
Informazioni di solubilità Solubility in water: practically insoluble. Other solubilities: soluble in alcohol, ether and chloroform
Peso formulazione 100.2
Percent Purity ≥99%
Grado Spectroscopy
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.