missing translation for 'onlineSavingsMsg'
Learn More

n-Pentylcyclohexane, 98%, Thermo Scientific Chemicals

Catalog Number p-7025691
Click to view available options
Quantità:
2 g
10 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
4292-92-6
C11H22
154.297
MFCD00019402
HLTMUYBTNSVOFY-UHFFFAOYSA-N
n-amylcyclohexane, cyclohexane, pentyl, 1-cyclohexylpentane, n-pentylcyclohexane, amylcyclohexane, cyclohexane, n-pentyl, pentyl-cyclohexane, n-amyl cyclohexane, acmc-209jru, pentane, 1-cyclohexyl
20284
pentylcyclohexane
CCCCCC1CCCCC1
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
4292-92-6
C11H22
154.297
MFCD00019402
HLTMUYBTNSVOFY-UHFFFAOYSA-N
n-amylcyclohexane, cyclohexane, pentyl, 1-cyclohexylpentane, n-pentylcyclohexane, amylcyclohexane, cyclohexane, n-pentyl, pentyl-cyclohexane, n-amyl cyclohexane, acmc-209jru, pentane, 1-cyclohexyl
20284
pentylcyclohexane
CCCCCC1CCCCC1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico n-Pentylcyclohexane
Punto di fusione -58°C
Densità 0.8
Punti di ebollizione 202°C
Punto d'infiammabilità 63°C (145°F)
Odore Mild
Indice di rifrazione 1.4445
Quantità 2 g
Numero UN UN3082
Peso formulazione 154.3
Percent Purity 98%
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'