Learn More
Macherey-Nagel™ Tappi a vite trasparenti in polipropilene N9
Seal vials with screw caps with a secure septum to receive needles. The Macherey-NagelTM N9 pollypropylene transparent screw caps come with a choice of septa.
Marca: Macherey-Nagel™ 702287
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Screw closures N 9
- Large range of screw closures with different septum materials and cap colors
- Bonded screw caps (septa firmly connected with the cap; cannot be removed) – advantage: thick (blunt) HPLC needles cannot push the septum into the vial
- Polymagnetic screw closure 702166 for CTC GC PAL
Specifica
Trasparente | |
Silicone bianco/PTFE rosso | |
N9 |
Polipropilene, PTFE, Silicone | |
1 mm |