missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ Color-Coded LDPE Wash Bottles
Click to view available options
Colore chiusura:
Blue
Giallo
Green
Rosso
Descrizione
High quality squeeze bottles with color-coded closures for easy identification. Thermo Scientific™ Nalgene™ Color-Coded LDPE Wash Bottles are made from lab quality low density polyethylene (LDPE) for dependably low leachables and chemical compatability with a wide range of lab reagents. Enjoy smooth, precision, sputter-free fluid dispensing with an ergonomic soft squeeze. Ideal for rinsing tubes and lab apparatus, bringing volumetric flasks to final volume and other precision rinsing and solution dispensing needs. The dispensing nozzle is molded in one piece with the bottle closure eliminating risk of separation when the bottle is squeezed. Cut back tip to increase flow.
- Color-coded top allows you to coordinate reagents with industry storage codes.
- Tip can be cut back to provide increased flow.
- Tight fit of draw tube and closure prevents separation when squeezing bottle.
Specifica
Specifica
Capacità (sistema metrico) | 500 mL |
Capacità (sistema imperiale) | 16 oz. |
Colore | Natural |
Descrizione | With stem and draw tube |
Materiale | LDPE |
N. per custodia | 24 |
N. per confezione | 6 |
Colore chiusura | Green |
Closure Included | Yes |
Autoclavabile | Not Autoclavable |
Vedi altri risultati |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'