Learn More
Thermo Scientific™ Nalgene™ Storm Water Samplers and Mounting Kit
Descrizione
The Nalgene™ Storm Water Sampler is a convenient and affordable device for collecting quarterly storm water grab samples in compliance with EPA sampling requirements. No more standing in the rain waiting for water to flow or missed sampling events. The Storm Water Sampler can collect a full one liter grab sample of first flush storm water runoff through a storm water ditch, stream or storm grate outfall. Simply position the sampler in its protective mounting tube prior to a rain event, and leave it in place until after the storm.
- UV-stabilized high-density polyethylene tube
- 24″ steel stake
- Includes stainless-steel worm clamp and lanyard, nylon rivets, polyester screen, wire hanger and carabiner-style snap
Specifica
Specifica
| Product Type | Mounting Kit |
| Descrizione | Storm Water Sampler Mount Kit |
| Include | HDPE Tube |
| Materiale provetta | Polyethylene |
| Certificazione/Conformità | Meets EPA NPDES MSGP (Fed. Reg. FRL-6880-5 10/20/00 and most state grab sampling requirements |