Learn More
Cytiva (formerly Pall Lab) Nanosep™ Centrifugal Devices with Omega™ Membrane
Use Nanosep™ devices to concentrate, purify, and desalt peptides, proteins, oligonucleotides, DNA, and RNA.
Brand: Cytiva (formerly Pall Lab) OD030C35
1961.75 EUR valido fino al 2024-12-31
Usa il codice promo "21930" per avere il tuo prezzo promozionale.
Additional Details : Peso : 1.32050kg
Description
- Simple, reliable,concentrating and desalting of 50 to 500 μL samples
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Red | |
PES | |
Non-sterile | |
500/Pk. |
EFA: 0.3 cm2 | |
30 kDa | |
Polypropylene | |
50 to 500 μL (Sample) |