missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NASP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11323-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
This antibody can recognize both isoforms.
Possesses single-stranded DNA-stimulated ATPase and ATP-dependent DNA helicase (3′ to 5′) activity. Component of the NuA4 histone acetyltransferase complex which is involved in transcriptional activation of select genes principally by acetylation of nucleosomal histones H4 and H2A. This modification may both alter nucleosome - DNA interactions and promote interaction of the modified histones with other proteins which positively regulate transcription. This complex may be required for the activation of transcriptional programs associated with oncogene and proto-oncogene mediated growth induction, tumor suppressor mediated growth arrest and replicative senescence, apoptosis, and DNA repair. The NuA4 complex ATPase and helicase activities seem to be, at least in part, contributed by the association of RUVBL1 and RUVBL2 with EP400. NuA4 may also play a direct role in DNA repair when recruited to sites of DNA damage. RUVBL1 plays an essential role in oncogenic transformation by MYC and also modulates transcriptional activation by the LEF1/TCF1-CTNNB1 complex. May be able to bind plasminogen at cell surface and enhance plasminogen activation. Essential for cell proliferation.Specifica
NASP | |
Polyclonal | |
Unconjugated | |
NASP | |
FLB7527, NASP, PRO1999 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
298441, 4678, 50927 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation, Immunocytochemistry | |
0.32 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P49321, Q66HD3, Q99MD9 | |
Nasp | |
NASP Fusion Protein Ag1824 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |