missing translation for 'onlineSavingsMsg'
Learn More

Naspm trihydrochloride, Tocris Bioscience™

Codice prodotto. 18055992
Click to view available options
Quantità:
10 mg
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Ca2+-permeable AMPA antagonist

Selective antagonist of Ca2+-permeable AMPA receptors; blocks AMPA receptors lacking the GluA2 subunit. Protects hippocampal neurons against global ischemia-induced cell death.

Specifica

Materiale o nome chimico Naspm trihydrochloride
CAS 1049731-36-3
Formula molecolare C22H37Cl3N4O
Quantità 10 mg
Sinonimo naspm trihydrochloride, 1-naphthylacetyl spermine trihydrochloride, n-3-4-3-aminopropyl amino butyl amino propyl-1-naphthaleneacetamide trihydrochloride, c22h34n4o.3clh, 1-naphthyl acetyl spermine trihydrochloride
InChI Key JNEOJAUJWOPWJS-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C=CC=C2CC(=O)NCCCNCCCCNCCCN.Cl.Cl.Cl
IUPAC Name N-[3-[4-(3-aminopropylamino)butylamino]propyl]-2-naphthalen-1-ylacetamide;trihydrochloride
Molecular Weight (g/mol) 479.915
PubChem CID 16219727
Peso formulazione 479.91
Percent Purity >97%
Vedi altri risultati Mostra meno risultati
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'