Learn More
BTX™ Needles Array Electrode Handles for BTX™ AgilePulse™ in Vivo
Needle Array Handle for miniature needles designed to provide superior, highly-uniform electric fields in dermal or muscular tissue as part of the AgilePulse™ in vivo Gene Delivery Electroporation System. Specially designed for gene and vaccine delivery.
Marca: BTX™ 470092
Descrizione
The miniature parallel-needle array is inserted directly into the target site for fast, reliable in vivo electroporation. The electric fields produced are the closest approximation of parallel plate electrodes, treating approximately 80% of the target area with 95% of the applied electric field.
The needle array requires the needle array handle which includes the electrode connector cable. Several needle lengths and row spacing are available for various intra-dermal or intra-muscular applications. Needle Arrays can be used for up to 500 pulses with proper care and maintenance.
- Uniform, reliable electric fields
- Miniature needles minimize tissue trauma
- Disposable and sterile
- Medical-grade plastic and surgical steel construction
- Safety-assured design
- Multiple configurations, sizes to choose from
Specifica
Gene and Vaccine Delivery | |
Plastic | |
Needle Array Handle | |
1 Ea. | |
Electrode Connector Cable |
Generators with SN AP-4069-ID/AP-5031-IM and After | |
BTX Electroporation | |
Handle Only | |
Non-sterile | |
Square Wave |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.