Learn More
Thermo Scientific Chemicals Nickel Cover for Crucible 35925, 50ml
Codice prodotto. 15411417
Click to view available options
Capacità (sistema metrico):
5 mL
20 mL
25 mL
30 mL
35 mL
45 mL
50 mL
55 mL
75 mL
100 mL
250 mL
500 mL
1000 mL
Descrizione:
For 35883 crucible
For 35889 crucible
For 35904 crucible
For 35906 crucible
For 35925 crucible
For 35927 crucible
For 35946 crucible
For 35969 crucible
For 35991 crucible
For 36019 crucible
For 36034 crucible
For 36040 crucible
For 36053 crucible
For 36057 crucible
For 36071 crucible
For 36084 crucible
Dimensione della confezione:
Pezzo
Codice prodotto. 15411417
Marca: Thermo Scientific Chemicals 036042.KT
Richiesta di formato massivo o personalizzato
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifica
Descrizione For 35906 crucible
Materiale Nickel
Capacità (sistema metrico) 20 mL
Product Type Nickel Crucible Cover