missing translation for 'onlineSavingsMsg'
Learn More

Nitrilotriacetic acid, trisodium salt, monohydrate, 99+%, Thermo Scientific Chemicals

Nitrilotriacetic acid, trisodium salt, monohydrate, CAS # 18662-53-8, is considered a non-genotoxic compound showing carcinogenic activity and is used as an inducer of tumorigenesis in laboratory studies using animal models.

28.44€ - 168.84€

Identificatori chimici

CAS 18662-53-8
Formula molecolare C6H6NNa3O6·H2O
Molecular Weight (g/mol) 275.09
InChI Key HERBOKBJKVUALN-UHFFFAOYSA-K
Sinonimo trisodium nitrilotriacetate monohydrate, nitrilotriacetic acid trisodium salt monohydrate, nta sodium hydrate, nta trisodium salt dihydrate, unii-j0u92u6nu5, ccris 437, nitrilotriacetic acid trisodium salt, monohydrate, nitriloacetic acid trisodium salt monohydrate, glycine, n,n-bis carboxymethyl-, trisodium salt, monohydrate, nitrilotriacetic acid, trisodium salt monohydrate
PubChem CID 29194
IUPAC Name trisodium;2-[bis(carboxylatomethyl)amino]acetate;hydrate
SMILES C(C(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].O.[Na+].[Na+].[Na+]
Visualizza altre specifiche

Prodotti 3
Codice del prodotto Marca Quantità Imballaggio Prezzo Quantità e disponibilità  
Codice del prodotto Marca Quantità Imballaggio Prezzo Quantità e disponibilità  
10498290
Vedere i documenti
Thermo Scientific Acros
147840250
25 g Plastic bottle
28.44€
25g
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
10408300
Vedere i documenti
Thermo Scientific Acros
147845000
500 g Plastic bottle
49.85€
500g
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
11985331
Vedere i documenti
Thermo Scientific Acros
147840025
2.5 kg Plastic bottle
168.84€
2.50kg
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
Descrizione

Descrizione

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Information

• Nitrilotriacetic acid, trisodium salt, monohydrate is considered to be a nongenotoxic compound with carcinogenic activity
• This compound inhibits gap-junctional communication between Chinese hamster lung fibroblasts

Applications

• Nitrilotriacetic acid trisodium salt monohydrate is commonly used in laboratory experiments due to its capacity to induce the tumorigenic process
• It has been used in in vivo experiments as an inducer of bladder and kidney tumors in animal models
• It is used as a chelating agent, which forms coordination compounds with metal ions (chelates)

Literature References
Malcolm, A.; Mills, L. Inhibition of gap-junctional intercellular communication between Chinese hamster lung fibroblasts by di(2-ethylhexyl) phthalate (DEHP) and trisodium nitrilotriacetate monohydrate (NTA). Cell Biology and Toxicology. 1989, 5, 145–153.

Specifica

Identificatori chimici

18662-53-8
275.09
trisodium nitrilotriacetate monohydrate, nitrilotriacetic acid trisodium salt monohydrate, nta sodium hydrate, nta trisodium salt dihydrate, unii-j0u92u6nu5, ccris 437, nitrilotriacetic acid trisodium salt, monohydrate, nitriloacetic acid trisodium salt monohydrate, glycine, n,n-bis carboxymethyl-, trisodium salt, monohydrate, nitrilotriacetic acid, trisodium salt monohydrate
trisodium;2-[bis(carboxylatomethyl)amino]acetate;hydrate
C6H6NNa3O6·H2O
HERBOKBJKVUALN-UHFFFAOYSA-K
29194
C(C(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].O.[Na+].[Na+].[Na+]

Specifica

Nitrilotriacetic acid, trisodium salt, monohydrate
>320.0°C
Authentic
C6H6NNa3O6·H2O
trisodium nitrilotriacetate monohydrate, nitrilotriacetic acid trisodium salt monohydrate, nta sodium hydrate, nta trisodium salt dihydrate, unii-j0u92u6nu5, ccris 437, nitrilotriacetic acid trisodium salt, monohydrate, nitriloacetic acid trisodium salt monohydrate, glycine, n,n-bis carboxymethyl-, trisodium salt, monohydrate, nitrilotriacetic acid, trisodium salt monohydrate
HERBOKBJKVUALN-UHFFFAOYSA-K
trisodium;2-[bis(carboxylatomethyl)amino]acetate;hydrate
29194
99+%
18662-53-8
White
99+%
N(CH2CO2Na)3·H2O
Solubility in water: 484g/L (25°C)
C(C(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].O.[Na+].[Na+].[Na+]
275.09
275.09
Powder, Crystals or Chunks
Videos
Sicurezza e movimentazione

Sicurezza e movimentazione

GHS H Statement
Suspected of causing cancer.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement
Use personal protective equipment as required.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present an

GHS Signal Word: Warning

SDS
Documenti

Documenti

Product Certifications

RUO – Research Use Only

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato