missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One Non-treated 384-well Microplates
Manufactured without surface treatment to create low biomolecular binding. Greiner Bio One™ Non-treated 384-Well Assay Microplates are ideal for sensitive biochemical screening applications.
Marca: Greiner Bio-One 788101
Descrizione
- Footprint compatible with automated systems
- Can be traced to production through defined LOT number system
- Free of detectable DNase, RNase and human DNA
- Non-pyrogenic
- Manufactured under DIN ISO 9001 guidelines
- Quadrupled the number of wells (from 96)
- Well-to-well spacing: 4.5 mm
- Rounded square well profile, providing flexible working volume of 10 μl to 130 μl and reduced wicking and bubbling
Specifica
DIN ISO 9001 | |
16 x 24 | |
Clear | |
384 | |
Non-treated | |
Flat Bottom (F-well) | |
80/Pk. |
Standard | |
Medium Binding | |
Polystyrene | |
Non-sterile | |
131 μL | |
80 |