Learn More
Thermo Scientific™ Novex™ Tricine XCell SureLock™ Mini-Cell Welcome Pack, 10-20%
Sfoglia Tutto Thermo Scientific ProdottiDescrizione
The Novex Tricine Welcome Pack provides all of the necessary Novex Tricine gels, buffers, and reagents you will need to begin using the XCell SureLock Mini-Cell. The XCell SureLock Mini-Cell is a vertical mini-protein gel electrophoresis system compatible with all Invitrogen mini precast gels and Surecast handcast gels.
The Novex Tricine Welcome Pack contains:
• XCell SureLock Mini-Cell (EI0001)
• Novex Tricine Mini Gels (2 boxes, 20 gels)
• Novex Tricine SDS Running Buffer, 10X (LC1675)
• Novex Tricine SDS Sample Buffer, 2X (LC1676)
• NuPAGE Sample Reducing Agent, 10X (NP0009)
• Spectra Multicolor Low Range Protein Ladder (26628)
About the XCell SureLock Mini-Cell
The XCell SureLock Mini-Cell is a vertical mini-protein gel electrophoresis system compatible with all Invitrogen mini precast gels and Surecast handcast gels. The Mini-Cell is simple, sturdy, and easy to use. Each XCell SureLock Mini-Cell can accommodate up to two gels per run. Run times may vary depending on buffer conditions and power supplies used.
About Novex Tricine gels
Novex Tricine gels provide separation of low molecular weight proteins and peptides. The Tricine system is a modification of the tris-glycine discontinuous buffer system developed by Schaegger and von Jagow specifically for peptides and low molecular weight proteins. In this system the tricine replaces the glycine in the running buffer, resulting in more efficient stacking and de-stacking of low molecular weight proteins and higher resolution of smaller peptides.
Transfer
For transfer of proteins to a membrane, we recommend using Novex Tris-Glycine Transfer Buffer (LC3675) for traditional wet transfer using the XCell II Blot Module (EI9051). Alternatively, rapid semi-dry transfer can be performed using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 3 Gel Transfer Device (IB31001).
Specifications
Specifications
| Da utilizzare con (apparecchiatura) | XCell SureLock Mini-Cell |
| Gel Percentage | 10 to 20% |
| Gel Size | Mini |
| Gel Thickness | 1.0 mm |
| Gel Type | Tricine |
| Separation Range | 2.5 to 200 kDa |
| Pozzetti | 12-well |
| Linea di prodotti | Novex |
| Quantità | 1 Welcome Pk. |
| Shipping Condition | Wet Ice |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.