Learn More
Novus Biologicals™ Recombinant Human p62/SQSTM1 His Protein
Codice prodotto. 18718453 Sfoglia Tutto Bio Techne Prodotti
Click to view available options
Quantità:
0.1 mg
Dimensione della confezione:
0.10mg
Codice prodotto. 18718453
Marca: Novus Biologicals™ NBP144490
Highly purified. Generating reliable and reproducible results. Applications: SDS-Page
A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 1-356 of Human p62/SQSTM1 The Recombinant Human p62/SQSTM1 Protein is derived from E. coli. The Recombinant Human p62/SQSTM1 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY
Specifica
Quantità 0.1 mg