Learn More
Thermo Scientific™ Nunc™ Lab-Tek™ II Chamber Slide™ System
Codice prodotto. 11377704
Click to view available options
Pozzetti:
1
2
4
8
Dimensione della confezione:
pezzi 16
pezzi 96
Codice prodotto. 11377704
Marca: Thermo Scientific™ 154461
Grow cells directly on slides with the Nunc Lab-Tek II Chamber Slide System.
Grow, stain, and examine cells directly on the Thermo Scientific Nunc Lab-Tek II Chamber Slide, which consists of a removable polystyrene media chamber attached to a standard glass slide. The slide surface is specially treated for adherent cell culture. Available in 1-, 2-, 4-, and 8-well formats.
Features
  • Slide surface that is specially treated with a unique RS wash to ensure consistent cell growth
  • Tabbed polystyrene lid that allows gas exchange with minimal evaporation
  • Removal tool that separates media chamber from glass slide
  • Biocompatible adhesive hydrophobic well border printed on slide
  • White frosted writing area
Includes: chamber removal tool
Additional information
  • Can be purchased in case of 96 or pack of 16
  • Shipped sterile in convenient stackable incubation trays
Warranty: 90 days
TRUSTED_SUSTAINABILITY
Specifica
Pozzetti 2
Area di coltura 4.0 cm2
Materiale del coperchio Polystyrene
Descrizione 2-well Chamber Slide w/ removable wells
Trattamento superficiale Cell Culture
Materiale RS Glass
Volume (sistema metrico) di lavorazione Da 1.0 a 2.0 mL
Include Clear polystyrene lid with handles on the top and side for aseptic manipulations
N. per confezione 16
Dimensions (LxW) 74 x 25 mm
Sterilità Sterile
Colore chiusura Clear
Quantità 1 Ea.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.