Learn More
Kolmi Hopen™ Op-Air™ One Type IIR Printed Medical Mask with Ties
This splash-resistant medical mask is formulated with a soft inner layer for sensitive skin.
Marca: Kolmi Hopen™ M77100-GEO
Dettagli aggiuntivi : Peso : 1.52000kg
Descrizione
- Protects environment from droplets emitted by wearer
- Feature exclusive Origami folding and Smile welding for better comfort and perfect fit
- Feeldry inner layer absorbs moisture to prevent skin reactions
- Intended for healthcare professionals
- May also be worn by patients to reduce risk of spreading infection
Specifica
Blue Geometric Print | |
Polypropylene | |
(EU) 2017/745, EN 14683:2019+AC:2019, ISO 9001, ISO 13485 | |
Yes |
Ties | |
175 x 95 mm | |
Yes | |
40 Pk. |