Learn More
Kolmi Hopen™ Op-Air™ Type II Medical Mask with Ties and Polypropylene Interior Layer
Tie-on medical masks for use in healthcare setting.
Marca: Kolmi Hopen™ M21101-30
6.07 EUR valido fino al 2024-09-30
Usa il codice promo "21675" per avere il tuo prezzo promozionale.
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- High filtration media (polypropylene)
- Layers (interior to exterior): thermobonded polypropylene, melt blown polypropylene, spunbonded polypropylene
- Soft, non-woven mask ties and edges
- Anatomical folding limits exposure to face
- Integrated nose bar
- Fiberglass-free filter
- Produced in ISO 8 clean room (particle and microbiological control)
Specifica
White | |
Polypropylene | |
(EU) 2017/745, EN 14683, ISO 13485, ISO 9001 | |
Medical |
Ties | |
Pleated | |
Single-use | |
50 Pk. |