Learn More
Köttermann Open Tall Storage Cabinet
The open-front design of this Open Tall Storage Cabinet provides easy access to stored equipment and utensils.
Marca: Köttermann 301-00080
Dettagli aggiuntivi : Peso : 75.80990kg
Descrizione
- Tall, slender design expands storage with a small footprint
- Open front provides easy access to stored items
Specifica
None | |
Storage | |
No | |
75.8 kg | |
4 | |
Epoxy Coated | |
White |
None | |
No | |
500 x 900 x 1920 mm | |
None | |
Tall Storage | |
None | |
Steel |