Learn More
Corning™ Organoid Counting Software for the Corning™ Cell Counter
Accurate – powered by machine learning artificial intelligence.
Marca: Corning™ 6749-OC
Dettagli aggiuntivi : Peso : 0.08000kg
Descrizione
The Organoid counting software ships with a 0.2 mm, 20 μL, counting chamber and an alphanumeric access code. This software extension enables Corning™ Cell Counter operators to capture rapid data of 3D cell cultures based on the structure’s morphology. Results are viewed within the intuitive application interface or remotely accessible by utilizing the cloud login. Unlimited storage.
- Accurate – powered by machine learning artificial intelligence
- Multi-informative – organoids per milliliter of solution and surface area data
- Fast – three-second analysis utilizing cloud-based processing
- Cost-efficient and versatile – compatible with standard 0.1 mm counting chambers and 0.2 mm counting chambers. No consumables required.
Specifica
Cell Counting Software | |
Corning™ Cell Counter | |
0.2 mm, 20 μL, counting chamber and an alphanumeric access code |