Learn More
Thermo Scientific™ Orion™ AQUAfast™ IV Chemistry Reagents
Descrizione
Minimize the risks of improper measurement and contamination with Thermo Scientific™ Orion™ AQUAfast™ IV Chemistry Reagents, which are contained in self-filling vials. Use these reagents with the Orion™ AQUAfast AQ4000 colorimeter to simplify sample preparation and tests. The auto-test ampoules contain a pre-measured amount of liquid reagent and easily fill with the correct volume of sample for simple and fast sample and reagent mixing. Each ampoule contains an Auto-ID barcode that is read by the AQ4000 colorimeter and used to automatically identify the test parameter and initiates the test.
- No. of test: 30
- Consistency and reproducibility improved with AQUAfast ampoules and an AQ4000 meter
- Thermo Scientific™ Auto-ID™ eliminates measurement errors
Specifica
Specifica
| Tipo | Sulfide |
| Formati reagenti test | Ampoule |
| Da utilizzare con (apparecchiatura) | Orion AQUAfast AQ4000 Colorimeter |
| Descrizione | AQUAfast sulfide (Acid Soluble) Auto-ID ampoules, 30 pack |
| Include | 30 Auto-ID Ampoules |
| Metodo | Methylene Blue |
| Da utilizzare con (applicazione) | Colorimetric water and wastewater testing equipment |
| Accessori raccomandati | AQUAfast Zero Vial Auto-Test Kit (Cat. No. AQ4ZER) |
| Range | 0.2 to 3 mg/L S |
| Metodi approvati EPA | Wastewater: Standard Methods 4500-S2- D |
| Vedi altri risultati |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.