Learn More
Thermo Scientific™ Orion™ Membrane Caps for Polarographic Dissolved Oxygen Probe
Reduce probe servicing time and eliminate assembly problems using the Thermo Scientific™ Orion™ Membrane Caps for Polarographic Dissolved Oxygen Probe.
Marca: Thermo Scientific™ 081003
Dettagli aggiuntivi : Peso : 0.80000kg
Descrizione
Reduce probe servicing time and eliminate assembly problems using Thermo Scientific™ Orion™ Membrane Caps for Polarographic Dissolved Oxygen Probe. Convenient screw-on membrane caps help ensure optimal probe performance.
Warranty: Out of boxSpecifica
Orion DO Probes 080510MD, 081010MD, 081010 | |
Dissolved Oxygen Probe Accessories | |
Three Caps | |
Polarographic DO |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.