Learn More
Thermo Scientific™ Orion™ Resistor Kits and Flow Cell Conductivity Probe Accessories
Perform virtually any electrochemistry analysis with these conductivity standards, solutions and accessories.
Marca:  Thermo Scientific™ 013017
 vedi altre versioni di questo prodotto
Codice prodotto. 17298959
Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Visualizzare prodotti alternativi »
Esplora altre offerte speciali
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
Confidently perform conductivity tests and measurements with Thermo Scientific™ Orion™ Accessories. Find an additional or replacement Glass Flow Cell for use with the Orion 2-Cell Ultra-pure Conductivity Probe. Use the Thermo Scientific Orion Conductivity Resistor Calibration Kit to verify the performance of Thermo Scientific™ Orion™ Versa Star Pro™ and Orion Star™ Bench and Portable Conductivity Meters. Each conductivity calibration kit includes six resistors that simulate nominal conductance values between 1 μS and 100 mS and have 8 pin MiniDIN connectors.
Conductivity Standards
  • Traceable to NIST standard reference materials
  • Designed to meet all end user requirements, including laboratory grade and DIN standards
  • Conductivity standard temperature tables are programmed into Thermo Scientific Orion meter software for use with the automatic calibration feature
TRUSTED_SUSTAINABILITY
Specifica
Specifica
Replacement Glass Flow Cell
Conductivity Probes (013016MD, 013016A and 013016D)
Replacement Flow Cell
Yes
Suggerimenti di prodotto
Suggerimenti di prodotto
Videos
SDS
Documenti
Documenti
Certificati
Offerte speciali
Offerte speciali
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.