Learn More
Osmometers
Marca: Loeser TYP15M
Questo articolo non è restituibile.
Consulta la politica di reso
Specifica
LCD Graphic | |
230 V | |
216 mm | |
Osmometer | |
Food: milk water content. Hospital: control injections. Transfusion Centre: control of physicochemical properties chemical derivatives of blood and analyzed for residual glycerol after thawing. Cell culture, biochemistry, marine biology, plant physiology | |
278 mm | |
230 VAC (100/115 VAC on request) approx. 45 VA, 50 Hz, 20/45 w | |
1 mOsm/kg | |
20/45 w | |
180 mm |
50 Hz | |
Peltier Effect | |
1 Ea. | |
Type 15M | |
Raised By Higher Concentrations | |
1.3 min. | |
0 to 2500 mosm/kg | |
10 to 50 μl | |
6.3 kg | |
230 VAC (100/115 VAC on request) approx. 45 VA, 50 Hz, 20/45 w |