Learn More
Thermo Scientific™ Oxoid™ Listeria Test Kit
Thermo Scientific™ Oxoid Listeria Kit is a rapid latex agglutination test used for presumptive identification of Listeria spp. in cultures.
Marca: Thermo Scientific™ DR1126A
Dettagli aggiuntivi : Peso : 0.50000kg
Descrizione
Thermo Scientific™ Oxoid Listeria Kit is a rapid latex agglutination test used for presumptive identification of Listeria spp. in cultures.
Microtiter plate is inoculated with enriched pre-tested food samples and washed with monoclonal antibody conjugate which binds to Salmonella antigens forming an immune complex. 8 breakaway strips.Specifica
Listeria Test Kit | |
Pre-Tested Food | |
50 Tests/Kit | |
Listeria Spp. |
Listeria species | |
Listeria Test Kit | |
50 Tests/Kit |